Strontium nitrate ≥99.995%

Warning

204498-50G 204498-10G
220042-905EA 793.85 USD
220042-905 220042-904
Strontium nitrate ≥99.995%
Strontium nitrate
Formula: Sr(NO₃)₂
MW: 211.63 g/mol
Boiling Pt: 645 °C (1013 hPa)
Melting Pt: 570 °C
Density: 2.113 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011248
CAS Number: 10042-76-9
EINECS: 233-131-9
UN: 1507
ADR: 5.1,III
Merck Index: 13,08926

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR