Strontium chloride hexahydrate ≥99.995%

Warning

204463-10G 204463-50G
220042-902EA 292.31 USD
220042-902 220042-903
Strontium chloride hexahydrate ≥99.995%
Strontium chloride hexahydrate
Formula: SrCl₂·6H₂O
Boiling Pt: 100 °C (1013 hPa)
Melting Pt: 61 °C
Density: 1.93 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149865
CAS Number: 10025-70-4
EINECS: 233-971-6
Merck Index: 13,08920

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR