Ammonium hexafluorosilicate

Danger

Synonyms: Ammonium silicofluoride Ammonium hexafluorosilicate, Ammonium fluorosilicate, Ammomium fluosilicate

204331-50G 204331-10G
220042-631EA 327.69 USD
220042-631 220042-629
Ammonium hexafluorosilicate
Ammonium hexafluorosilicate
Formula: H₈F₆N₂Si
MW: 178.15 g/mol
Density: 2.01 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010887
CAS Number: 16919-19-0
EINECS: 240-968-3
UN: 2854
ADR: 6.1,III
Merck Index: 13,00529

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR