Praseodymium(III,IV) oxide ≥99.999%

Warning

204145-10G 204145-2G
220042-891EA 1406.15 USD
220042-891 220041-947
Praseodymium(III,IV) oxide ≥99.999%
Praseodymium(III,IV) oxide
Formula: O₁₁Pr₆
MW: 1021.44 g/mol
Melting Pt: 2012…2072 °C
Storage Temperature: Ambient
MDL Number: MFCD00011178
CAS Number: 12037-29-5
EINECS: 234-857-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR