Nickel(II) nitrate hexahydrate

Danger

203874-100G 203874-20G 203874-500G
220042-845EA 324.62 USD
220042-845 220042-846 220042-847
Nickel(II) nitrate hexahydrate
Nickel(II) nitrate hexahydrate
Formula: Ni(NO₃)₂·6H₂O
Boiling Pt: 137 °C (1013 hPa)
Melting Pt: 56.7 °C
Density: 2.05 g/cm³ (20 °C)
MDL Number: MFCD00149805
CAS Number: 13478-00-7
EINECS: 236-068-5
UN: 2725
ADR: 5.1,III
Merck Index: 12,06596

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR