Nickel(II) chloride hexahydrate

Danger

Synonyms: Nickel dichloride hexahydrate

203866-25G 203866-5G
220042-843EA 284.62 USD
220042-843 220042-844
Nickel(II) chloride hexahydrate
Nickel(II) chloride hexahydrate
Formula: NiCl₂·6H₂O
MW: 237.69 g/mol
Melting Pt: 140 °C
Density: 1.84 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149809
CAS Number: 7791-20-0
EINECS: 231-743-0
UN: 3288
ADR: 6.1,III
Merck Index: 13,06529

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR