Manganese(II) nitrate hydrate

Warning

Synonyms: Manganous nitrate hydrate

203742-100G 203742-25G
220042-332EA 578.46 USD
220042-332 220042-333
Manganese(II) nitrate hydrate
Manganese(II) nitrate hydrate
Formula: Mn(NO₃)₂·H₂O
MW: 196.96 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00149788
CAS Number: 15710-66-4
EINECS: 233-828-8
UN: 2724
ADR: 5.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR