Manganese(II) chloride tetrahydrate

Danger

Synonyms: Manganese dichloride tetrahydrate, Manganese chloride (tetrahydrate), Manganese(II) chloride tetrahydrate, Manganous chloride tetrahydrate, Manganese chloride tetrahydrate, Manganese Chloride, Tetrahydrate

203734-100G 203734-25G 203734-5G
220042-329EA 394.46 USD
220042-329 220042-330 220042-331
Manganese(II) chloride tetrahydrate
Manganese(II) chloride tetrahydrate
Formula: MnCl₂·4H₂O
MW: 197.9 g/mol
Boiling Pt: 1190 °C (1013 hPa)
Melting Pt: 58…59 °C
Density: 2.01 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149792
CAS Number: 13446-34-9
EINECS: 231-869-6
UN: 3077
ADR: 9,III
Merck Index: 13,05751

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR