Lithium chloride ≥99.99%

Warning

203637-10G 203637-50G 203637-100G
220042-835EA 144.15 USD
220042-835 220042-836 220042-834
Lithium chloride ≥99.99%
Lithium chloride
Formula: LiCl
MW: 42.39 g/mol
Boiling Pt: 1382 °C (1013 hPa)
Melting Pt: 614 °C
Density: 2.07 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011078
CAS Number: 7447-41-8
EINECS: 231-212-3
Merck Index: 13,05550

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR