Lead(II) oxide ≥99.999%

Danger

203610-10G 203610-50G
220042-830EA 142.62 USD
220042-830 220042-831
Lead(II) oxide ≥99.999%
Lead(II) oxide
Formula: PbO
Boiling Pt: 1470 °C (1013 hPa)
Melting Pt: 888 °C
Density: 9.5 g/cm³ (20 °C)
MDL Number: MFCD00011164
CAS Number: 1317-36-8
EINECS: 215-267-0
UN: 2291
ADR: 6.1,III
Merck Index: 13,05431

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR