Lanthanum(III) nitrate hexahydrate ≥99.999%

Warning

203548-100G 203548-25G 203548-500G
220042-821EA 373.85 USD
220042-821 220042-822 220042-823
Lanthanum(III) nitrate hexahydrate ≥99.999%
Lanthanum(III) nitrate hexahydrate
Formula: La(NO₃)₃·6H₂O
MW: 433.01 g/mol
Boiling Pt: 126 °C (1013 hPa)
Melting Pt: 65…67 °C
Density: 2.347 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149751
CAS Number: 10277-43-7
EINECS: 233-238-0
UN: 1477
ADR: 5.1,III
Merck Index: 14,05363

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR