Lanthanum(III) chloride heptahydrate ≥99.999%

203521-100G 203521-25G
220042-819EA 520 USD
220042-819 220042-820
Lanthanum(III) chloride heptahydrate ≥99.999%
Lanthanum(III) chloride heptahydrate
Formula: LaCl₃·7H₂O
MW: 371.37 g/mol
Boiling Pt: ∼860 °C (1013 hPa)
Melting Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00149756
CAS Number: 10025-84-0
EINECS: 233-237-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR