Indium(III) chloride ≥99.999%

Synonyms: Indium Trichloride Chloride, Indium trichloride

203440-1G 203440-10G 203440-50G
220042-806EA 66.15 USD
220042-806 220042-805 220042-807
Indium(III) chloride ≥99.999%
Indium(III) chloride
Formula: Cl₃In
MW: 221.18 g/mol
Boiling Pt: 800 °C (1013 hPa)
Melting Pt: 586 °C
Density: 4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011058
CAS Number: 10025-82-8
EINECS: 233-043-0
UN: 3260
ADR: 8,II
Merck Index: 13,04980

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR