Indium ≥99.999%

203432-25G 203432-5G
220042-803EA 1569.23 USD
220042-803 220042-804
Indium ≥99.999%
Indium
Formula: In
MW: 114.82 g/mol
Density: 7.31 g/cm³ (20 °C)
MDL Number: MFCD00134048
CAS Number: 7440-74-6
EINECS: 231-180-0
UN: 3089
ADR: 4.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR