Germanium ≥99.99%

203351-50G 203351-10G
220042-796EA 1569.23 USD
220042-796 220042-795
Germanium ≥99.99%
Germanium
Formula: Ge
MW: 72.61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00085310
CAS Number: 7440-56-4
EINECS: 231-164-3
UN: 3089
ADR: 4.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR