Gadolinium trichloride hexahydrate ≥99.999%

Warning

Synonyms: Gadolinium trichloride hexahydrate

203289-5G 203289-1G 203289-25G
220042-745EA 233.85 USD
220042-745 220042-743 220042-744
Gadolinium trichloride hexahydrate ≥99.999%
Gadolinium trichloride hexahydrate
Formula: Cl₃Gd·6H₂O
MW: 371.7 g/mol
MDL Number: MFCD00149732
CAS Number: 13450-84-5
EINECS: 233-386-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR