Europium(III) oxide ≥99.99%

203262-25G 203262-5G
220042-741EA 1769.23 USD
220042-741 220042-742
Europium(III) oxide ≥99.99%
Europium(III) oxide
Formula: Eu₂O₃
MW: 351.92 g/mol
Melting Pt: 2325 °C
Density: 7.42 g/cm³ (20 °C)
MDL Number: MFCD00010997
CAS Number: 1308-96-9
EINECS: 215-165-6
Merck Index: 14,03904

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR