Europium(III) chloride hexahydrate ≥99.99%

Synonyms: Europium(III) chloride hydrate, Europium trichloride hexahydrate

203254-5G 203254-25G 203254-1G
220042-740EA 258.46 USD
220042-740 220042-739 220042-241
Europium(III) chloride hexahydrate ≥99.99%
Europium(III) chloride hexahydrate
Formula: Cl₃Eu·6H₂O
MW: 366.41 g/mol
Melting Pt: 158 °C
Density: 4.89 g/cm³ (20 °C)
MDL Number: MFCD00010994
CAS Number: 13759-92-7
EINECS: 233-040-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR