Erbium(III) oxide ≥99.99%

Warning

Synonyms: Dierbium trioxide, Erbium oxide

203238-25G 203238-5G
220041-764EA 566.15 USD
220041-764 220041-765
Erbium(III) oxide ≥99.99%
Erbium(III) oxide
Formula: Er₂O₃
MW: 382.52 g/mol
Melting Pt: 2420 °C
Density: 7.36 g/cm³ (20 °C)
MDL Number: MFCD00010991
CAS Number: 12061-16-4
EINECS: 235-045-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR