Dysprosium(III) oxide ≥99.99%

Synonyms: Didysprosium trioxide, Dysprosium oxide

203181-5G 203181-25G
220041-756EA 158.46 USD
220041-756 220041-755
Dysprosium(III) oxide ≥99.99%
Dysprosium(III) oxide
Formula: Dy₂O₃
MW: 373 g/mol
Melting Pt: ~ 2400 °C
Density: 7.81 g/cm³ (20 °C)
MDL Number: MFCD00010986
CAS Number: 1308-87-8
EINECS: 215-164-0
Merck Index: 14,03482

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR