Copper(II) sulfate pentahydrate

Warning

Synonyms: Cupric sulphate pentahydrate, Copper sulphate pentahydrate

203165-50G 203165-10G
220041-753EA 430.77 USD
220041-753 220041-752
Copper(II) sulfate pentahydrate
Copper(II) sulphate pentahydrate
Formula: CuSO₄·5H₂O
MW: 249.69 g/mol
Density: 2.284 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149681
CAS Number: 7758-99-8
EINECS: 231-847-6
UN: 3077
ADR: 9,III
REACH: 01-2119520566-40
Merck Index: 13,02682

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR