Chromium(III) oxide ≥99.9% (trace metals basis), powder, Sigma-Aldrich®

Synonyms: Chromic oxide

203068-5G 203068-25G
220042-725EA 49.11 USD
220042-725 220042-724
Chromium(III) oxide ≥99.9% (trace metals basis), powder, Sigma-Aldrich®
Chromium(III) oxide
Formula: Cr₂O₃
MW: 151.99 g/mol
Boiling Pt: 4000 °C (1013 hPa)
Melting Pt: 2435 °C
Density: 5.22 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010949
CAS Number: 1308-38-9
EINECS: 215-160-9
Merck Index: 13,02247

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR