Cesium chloride ≥99.999%

Synonyms: Cesium Chloride

203025-50G 203025-10G
220042-719EA 689.23 USD
220042-719 220042-718
Cesium chloride ≥99.999%
Cesium chloride
Formula: CsCl
MW: 168.36 g/mol
Boiling Pt: 1303 °C (1013 hPa)
Melting Pt: 646 °C
Density: 3.7808 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010955
CAS Number: 7647-17-8
EINECS: 231-600-2
Merck Index: 13,02021

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR