Cerium trinitrate hexahydrate ≥99.999%

Danger

Synonyms: Cerous chloride hexahydrate, Cerium (III) nitrate hexahydrate

202991-125G 202991-25G
220042-369EA 1320 USD
220042-369 220042-370
Cerium trinitrate hexahydrate ≥99.999%
Cerium trinitrate hexahydrate
Formula: Ce(NO₃)₃·6H₂O
MW: 434.22 g/mol
Boiling Pt: 200 °C (1013 hPa)
Melting Pt: 39 °C
Density: 2.377 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149631
CAS Number: 10294-41-4
EINECS: 233-297-2
UN: 1477
ADR: 5.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR