Orthoboric acid ≥99.999%

Danger

Synonyms: Boron trihydroxide, Boron hydroxide, Boric acid, Boracic acid

202878-250G 202878-50G 202878-10G
220042-703EA 551.85 USD
220042-703 220042-704 220042-702
Orthoboric acid ≥99.999%
Orthoboric acid
Formula: B(OH)₃
MW: 61.83 g/mol
Boiling Pt: 185 °C (1013 hPa)
Melting Pt: 160 °C
Density: 1.44 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011337
CAS Number: 10043-35-3
EINECS: 233-139-2
REACH: 01-2119486683-25
Merck Index: 13,01326
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=098dd700154daaf6338ebb270902bb2bc704f27d&vid=6eec7d529090dd4c4b966ed8cd8d057153c5a47b&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR