Bismuth(III) oxide ≥99.999%

Warning

Synonyms: Bismuth Trioxide, Dibismuth trioxide

202827-250G 202827-50G 202827-10G
220042-697EA 755.38 USD
220042-697 220042-698 220042-696
Bismuth(III) oxide ≥99.999%
Bismuth(III) oxide
Formula: BiO₃
MW: 465.96 g/mol
Boiling Pt: 1890 °C (1013 hPa)
Melting Pt: 825 °C
Density: 8.9 g/cm³ (20 °C)
MDL Number: MFCD00003462
CAS Number: 1304-76-3
EINECS: 215-134-7
Merck Index: 13,01273

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR