Aluminium sulfate ≥99.99%

Danger

Synonyms: Aluminum sulphate

202614-100G 202614-25G 202614-5G
220041-938EA 1356.92 USD
220041-938 220041-939 220041-940
Aluminium sulfate ≥99.99%
Aluminium sulphate
Formula: Al₂O₁₂S₃
MW: 342.15 g/mol
Melting Pt: 770 °C
Density: 2.672 g/cm³ (22.5 °C)
MDL Number: MFCD00003423
CAS Number: 10043-01-3
EINECS: 233-135-0
Merck Index: 12,00381

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR