Polyethylene glycol 400

Synonyms: PEG 400

202398-250G 202398-500G 202398-20KG 202398-5G
220042-593EA 83.29 USD
220042-593 220042-594 220042-592 220042-595
Polyethylene glycol 400
Polyethylene glycol 400
Formula: H(OCH₂CH₂)nOH
MW: 400 g/mol
Boiling Pt: ∼250 °C (1013 hPa)
Melting Pt: 4…8 °C
Density: 1.13 g/cm³ (20 °C)
Flash Pt: 245 °C
Storage Temperature: Ambient
MDL Number: MFCD00081839
CAS Number: 25322-68-3
EINECS: 500-038-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR