Polyethylene glycol 300

Synonyms: PEG 300

202371-5G 202371-20KG 202371-500G 202371-1KG 202371-250G
220041-590EA 57.14 USD
220041-590 220041-587 220041-589 220041-586 220041-588
Polyethylene glycol 300
Polyethylene glycol 300
Formula: H(OCH₂CH₂)nOH
MW: 300 g/mol
Melting Pt: –15…–10 °C
Density: 1.13 g/cm³ (20 °C)
Flash Pt: 220 °C
Storage Temperature: Ambient
MDL Number: MFCD00081839
CAS Number: 25322-68-3
EINECS: 500-038-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR