Polypropylene glycol, M.W. ∼4,000

Synonyms: Poly(propylene glycol), Poly(propylene oxide), PPG

202355-500G 202355-250G
220041-584EA 168.69 USD
220041-584 220041-583
Polypropylene glycol, M.W. ∼4,000
Polypropylene glycol
Formula: H(OCH₂CH₂CH₂)nOH
Storage Temperature: Ambient
MDL Number: MFCD00084446
CAS Number: 25322-69-4
EINECS: 500-039-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR