Sodium tetrafluoroborate ≥98%

202215-500G 202215-10KG 202215-25G
220042-588EA 54.15 USD
220042-588 220042-586 220042-587
Sodium tetrafluoroborate ≥98%
Sodium tetrafluoroborate
Formula: BF₄Na
Melting Pt: 384 °C
Density: ~ 2.47 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003515
CAS Number: 13755-29-8
EINECS: 237-340-6
Merck Index: 12,08761

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR