Cesium carbonate ≥99.9%

Warning

202126-500G 202126-25G 202126-5G 202126-100G
220042-565EA 420.23 USD
220042-565 220042-564 220042-566 220042-563
Cesium carbonate ≥99.9%
Cesium carbonate
Formula: Cs₂CO₃
MW: 325.82 g/mol
Melting Pt: 610 °C
Density: 4.24 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010957
CAS Number: 534-17-8
EINECS: 208-591-9
Merck Index: 13,02020

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR