Hafnium dioxide

202118-25G 202118-100G
220042-561EA 141.08 USD
220042-561 220042-560
Hafnium dioxide
Hafnium dioxide
Formula: HfO₂
MW: 210.49 g/mol
Boiling Pt: 5400 °C (1013 hPa)
Melting Pt: 2812 °C
Density: 9.68 g/cm³ (20 °C)
MDL Number: MFCD00003565
CAS Number: 12055-23-1
EINECS: 235-013-2
Merck Index: 14,04727

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR