Quinaldine red

Synonyms: 2-(2-(4-Dimethylamino)styryl)-1-ethylquinolinium iodide, 2-(4-(Dimethylamino)styryl)-1-ethylquinolin-1-ium iodide

201316-25G 201316-5G
220042-532EA 270.2 USD
220042-532 220042-533
Quinaldine red
Quinaldine red
Formula: C₂₁H₂₃IN₂
MW: 430.33 g/mol
Melting Pt: 240 °C
MDL Number: MFCD00011968
CAS Number: 117-92-0
EINECS: 204-221-5
Merck Index: 12,08230
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d4684321b9d88857feec4f61191f7654f87da7e&vid=13e692a42373b178b1b528f9de5c4b74b89d8a42&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR