Ammonium hexafluorophosphate

201138-500G 201138-25G 201138-100G
220042-509EA 510.92 USD
220042-509 220042-508 220042-507
Ammonium hexafluorophosphate
Ammonium hexafluorophosphate
Formula: H₄F₆NP
Melting Pt: 198 °C
Density: 2.18 g/cm³ (18 °C)
MDL Number: MFCD00064642
CAS Number: 16941-11-0
EINECS: 241-009-1
UN: 3260
ADR: 8,III
Merck Index: 12,00557

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR