Lithium hydride ≥95%

201049-5G 201049-100G 201049-10G 201049-500G
220042-436EA 77.54 USD
220042-436 220042-433 220042-434 220042-435
Lithium hydride ≥95%
Lithium hydride
Formula: HLi
MW: 7.95 g/mol
Melting Pt: 680 °C
Density: 0.685 to 0.725 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011074
CAS Number: 7580-67-8
EINECS: 231-484-3
UN: 1414
ADR: 4.3,I
Merck Index: 13,05555

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR