Potassium hexafluorophosphate ≥98%

Danger

200913-1KG 200913-50G 200913-250G
220041-773EA 592.31 USD
220041-773 220041-775 220041-774
Potassium hexafluorophosphate ≥98%
Potassium hexafluorophosphate
Formula: F₆KP
Melting Pt: 575 °C
Density: 2.58 g/cm³ (20 °C)
MDL Number: MFCD00011412
CAS Number: 17084-13-8
EINECS: 241-143-0
UN: 3260
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR