2-tert-Butyl-4-methoxyphenol

Synonyms: 3-BHA, 4-Hydroxy-3-tert-butylanisole, 3-tert-Butyl-4-hydroxyanisole

20021-100G 20021-500G
220042-266EA 88.31 USD
220042-266 220042-267
2-tert-Butyl-4-methoxyphenol
2-tert-Butyl-4-methoxyphenol
Formula: C₁₁H₁₆O₂
MW: 180.25 g/mol
Boiling Pt: 267…269 °C (1013 hPa)
Melting Pt: 60…61 °C
Density: 1.0587 g/cm³ (20 °C)
MDL Number: MFCD00040484
CAS Number: 121-00-6
EINECS: 204-442-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR