Selenium dioxide ≥99.9%

Danger

200107-5G 200107-25G
220042-252EA 35.78 USD
220042-252 220042-251
Selenium dioxide ≥99.9%
Selenium dioxide
Formula: SeO₂
MW: 110.96 g/mol
Melting Pt: 315 °C
Density: 3.95 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003562
CAS Number: 7446-08-4
EINECS: 231-194-7
UN: 3283
ADR: 6.1,II
Merck Index: 13,08508

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR