Lanthanum(III) oxide ≥99.99%

199923-500G 199923-100G
220042-192EA 872.31 USD
220042-192 220042-191
Lanthanum(III) oxide ≥99.99%
Lanthanum(III) oxide
Formula: La₂O₃
MW: 325.81 g/mol
Boiling Pt: 4200 °C (1013 hPa)
Melting Pt: 2315 °C
Density: 6.51 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011071
CAS Number: 1312-81-8
EINECS: 215-200-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR