4-Methylimidazole

Synonyms: 4-Methyl-1H-imidazole

199885-250G 199885-50G
220042-185EA 163.69 USD
220042-185 220042-186
4-Methylimidazole
4-Methylimidazole
Formula: C₄H₆N₂
MW: 82.11 g/mol
Boiling Pt: 263 °C (1013 hPa)
Density: 1.0416 g/cm³ (14.3 °C)
MDL Number: MFCD00005201
CAS Number: 822-36-6
EINECS: 212-497-3
UN: 3263
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR