Germanium(IV) oxide ≥99.998%

Warning

Synonyms: Germanium dioxide

199478-10G 199478-100G 199478-25G
220041-902EA 212.31 USD
220041-902 220041-901 220041-903
Germanium(IV) oxide ≥99.998%
Germanium(IV) oxide
Formula: GeO₂
MW: 104.61 g/mol
Melting Pt: 1110 °C
Density: 3.64 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011030
CAS Number: 1310-53-8
EINECS: 215-180-8
Merck Index: 12,04417

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR