(2-Fluorophenyl)acetonitrile ≥97%

Synonyms: 2-Fluorobenzyl cyanide, 2-(2-Fluorophenyl)acetonitrile

199346-50G 199346-10G
220042-171EA 155.08 USD
220042-171 220042-170
(2-Fluorophenyl)acetonitrile ≥97%
(2-Fluorophenyl)acetonitrile
Formula: C₈H₆FN
MW: 135.14 g/mol
Boiling Pt: 114 °C (1013 hPa)
Density: 1.14045 g/cm³ (15 °C)
MDL Number: MFCD00001897
CAS Number: 326-62-5
EINECS: 206-313-0
UN: 3276
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR