L(+)-Lactic acid ≥85% in aqueous solution

Danger

Synonyms: (S)-Lactic acid, L-Lactic acid, (S)-2-Hydroxypropanoic acid, (S)-(+)-2-Hydroxypropanoic acid, (S)-(+)-2-Hydroxypropionic acid, (S)-(+)-Lactic acid

199257-5G 199257-250G 199257-1KG
220042-161EA 43.62 USD
220042-161 220042-160 220042-159
L(+)-Lactic acid ≥85% in aqueous solution
L(+)-Lactic acid
Formula: C₃H₆O₃
MW: 90.08 g/mol
Density: 1.21 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00064266
CAS Number: 79-33-4
EINECS: 201-196-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR