2-(3-Hydroxyphenyl)ethanol ≥99%

Synonyms: 3-Hydroxyphenethyl alcohol

199028-10G 199028-1G
220042-139EA 695.77 USD
220042-139 220042-140
2-(3-Hydroxyphenyl)ethanol ≥99%
2-(3-Hydroxyphenyl)ethanol
Formula: C₈H₁₀O₂
MW: 138.17 g/mol
Boiling Pt: 124 - 126 °C (0.2 -rr)
Density: 1.082 g/cm³ (20 °C)
CAS Number: 13398-94-2
EINECS: 236-485-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR