Cesium fluoride ≥99%

Danger

198323-100G 198323-25G
220042-120EA 149.69 USD
220042-120 220042-121
Cesium fluoride ≥99%
Cesium fluoride
Formula: CsF
MW: 151.9 g/mol
Boiling Pt: 1251 °C (1013 hPa)
Melting Pt: 680 °C
Density: 4.627 g/cm³ (20 °C)
MDL Number: MFCD00010960
CAS Number: 13400-13-0
EINECS: 236-487-3
UN: 2923
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR