Toluidine blue O, certified by the Biological Stain Commission

Synonyms: TBO, 3-Amino-7-(dimethylamino)-2-methyl-5-phenothiazinium chloride, 3-Amino-7-(dimethylamino)-2-methylphenothiazin-5-ium chloride, Tolonium chloride, Basic Blue 17, Blutene chloride

198161-5G 198161-25G
220042-106EA 145.6 USD
220042-106 220042-105
Toluidine blue O, certified by the Biological Stain Commission
Toluidine blue O
Formula: C₁₅H₁₆ClN₃S
MW: 305.83 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011934
CAS Number: 92-31-9
EINECS: 202-146-2
Merck Index: 12,09658
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=913e9279ecf42a7d9996fe54fe196cfbe93cdd3c&vid=61820c198c4a39bab8f548d881a9f685e92f800a&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR