Sudan IV

Warning

Synonyms: Oil Red, 1-((2-Methyl-4-(o-tolyldiazenyl)phenyl)diazenyl)naphthalen-2-ol, Scarlet Red, Oil Red BB, 1-({2-Methyl-4-[(2-methylphenyl)diazenyl]-phenyl}diazenyl)-2-naphthol, Scarlet R, Biebrich scarlet R, Lipid Crimson, Scarlet Red Scharlach, Oil Red IV, Fat Red B, Solvent Red 24, 1-(2-Methyl-4-(2-methylphenylazo)phenylazo)-2-naphthol

198102-100G 198102-25G
220041-289EA 179.2 USD
220041-289 220042-102
Sudan IV
Sudan IV
Formula: C₂₄H₂₀N₄O
MW: 380.45 g/mol
Melting Pt: 188 °C
Storage Temperature: Ambient
MDL Number: MFCD00003893
CAS Number: 85-83-6
EINECS: 201-635-8
Merck Index: 12,08538
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=10ce9dd08992a21e0526a047ce197da922c95a7c&vid=a97ddaded4f6555d1480b2694694fca344d16c09&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR