2-Chloro-1-methylpyridinium iodide

198005-10G 198005-100G 198005-25G
220041-275EA 39.09 USD
220041-275 220041-274 220041-276
2-Chloro-1-methylpyridinium iodide
2-Chloro-1-methylpyridinium iodide
Formula: C₆H₇ClIN
MW: 255.49 g/mol
Melting Pt: 207 °C
MDL Number: MFCD00011984
CAS Number: 14338-32-0
EINECS: 238-288-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR