Quinuclidine

Synonyms: 1-Azabicyclo[2.2.2]octane, ABCO

197602-1G 197602-10G
220041-270EA 163.69 USD
220041-270 220042-101
Quinuclidine
Quinuclidine
Formula: C₇H₁₃N
MW: 111.19 g/mol
Boiling Pt: 150 °C (1013 hPa)
Density: 1.025 g/cm³ (20 °C)
MDL Number: MFCD00006690
CAS Number: 100-76-5
EINECS: 202-887-1
UN: 2811
ADR: 6.1,II
Merck Index: 12,08265

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR